Product Details

SNP ID
rs11605562
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:934594 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCCTAAGCTTTGGTGTCTGTCTA[C/T]GGGTTTGCTTAGAAGTGTAGAGACC
Phenotype
MIM: 607242
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP2A2 PubMed Links

Gene Details

Gene
AP2A2
Gene Name
adaptor related protein complex 2 alpha 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242837.1 Intron NP_001229766.1
NM_012305.3 Intron NP_036437.1
XM_011519929.1 Intron XP_011518231.1
XM_011519930.2 Intron XP_011518232.1

View Full Product Details