Product Details

SNP ID
rs3810794
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:2462522 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GACTGCGCATGAGGGAGGAGCTCTC[A/G]GCAGGTCAGTGGGTCCTGGGTCTGT
Phenotype
MIM: 600227 MIM: 602349
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C16orf59 PubMed Links

Gene Details

Gene
C16orf59
Gene Name
chromosome 16 open reading frame 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025108.2 793 Silent Mutation TCA,TCG S286S NP_079384.2
XM_011522667.1 793 Silent Mutation TCA,TCG S253S XP_011520969.1
Gene
CCNF
Gene Name
cyclin F
There are no transcripts associated with this gene.

Gene
LOC729652
Gene Name
uncharacterized LOC729652
There are no transcripts associated with this gene.

Gene
MIR6768
Gene Name
microRNA 6768
There are no transcripts associated with this gene.

Gene
NTN3
Gene Name
netrin 3
There are no transcripts associated with this gene.

View Full Product Details