Product Details

SNP ID
rs7211859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47908916 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCACCCCATCTCAACATCCTGACT[A/C]CAGCAGGGACACATGTCCTGCCTCT
Phenotype
MIM: 601801
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SP2 PubMed Links

Gene Details

Gene
SP2
Gene Name
Sp2 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003110.5 Intron NP_003101.3
XM_006722023.2 Intron XP_006722086.1
XM_006722025.2 Intron XP_006722088.1
XM_006722026.2 Intron XP_006722089.1
XM_006722027.2 Intron XP_006722090.1
XM_011525136.2 Intron XP_011523438.1
XM_011525137.2 Intron XP_011523439.1
XM_011525138.2 Intron XP_011523440.1
XM_011525139.2 Intron XP_011523441.1
XM_011525140.2 Intron XP_011523442.1
XM_011525142.1 Intron XP_011523444.1
XM_011525143.1 Intron XP_011523445.1
XM_017024968.1 Intron XP_016880457.1
XM_017024969.1 Intron XP_016880458.1
Gene
SP2-AS1
Gene Name
SP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details