Product Details

SNP ID
rs17507821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:174563911 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGGAAGGGAGTTAGGAGAAAACAC[C/T]GAAAGAAGCACCTCTTCCTCTCACC
Phenotype
MIM: 602357
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WIPF1 PubMed Links

Gene Details

Gene
WIPF1
Gene Name
WAS/WASL interacting protein family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077269.1 Intron NP_001070737.1
NM_003387.4 Intron NP_003378.3
XM_011511780.2 Intron XP_011510082.1

View Full Product Details