Product Details

SNP ID
rs34002881
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:39118659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCCATGTGCATACCTATATGCCA[A/G]GTGTCTTAGGAGATTTAAAAAAAAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM32 PubMed Links

Gene Details

Gene
ADAM32
Gene Name
ADAM metallopeptidase domain 32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001313994.1 Intron NP_001300923.1
NM_145004.6 Intron NP_659441.3
XM_006716298.2 Intron XP_006716361.1
XM_006716299.1 Intron XP_006716362.1
XM_011544433.1 Intron XP_011542735.1
XM_011544434.1 Intron XP_011542736.1
XM_017013183.1 Intron XP_016868672.1
XM_017013184.1 Intron XP_016868673.1
XM_017013185.1 Intron XP_016868674.1
XM_017013186.1 Intron XP_016868675.1
XM_017013187.1 Intron XP_016868676.1
XM_017013188.1 Intron XP_016868677.1

View Full Product Details