Product Details

SNP ID
rs12977517
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:49454380 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCACAATACCCTCACGGGGCTCCT[A/G]TGATCTCTCTAGCTCTCAGAATGTC
Phenotype
MIM: 613358 MIM: 611480
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH16A1 PubMed Links

Gene Details

Gene
ALDH16A1
Gene Name
aldehyde dehydrogenase 16 family member A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145396.1 Intron NP_001138868.1
NM_153329.3 Intron NP_699160.2
XM_011526441.1 Intron XP_011524743.1
XM_011526442.1 Intron XP_011524744.1
Gene
PIH1D1
Gene Name
PIH1 domain containing 1
There are no transcripts associated with this gene.

View Full Product Details