Product Details

SNP ID
rs11642892
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56731774 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCAGGTCACCACCTCTGGTTTGG[A/T]TTATTGAAATAGTTTCCCATTTGGT
Phenotype
MIM: 614351
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NUP93 PubMed Links

Gene Details

Gene
NUP93
Gene Name
nucleoporin 93
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242795.1 Intron NP_001229724.1
NM_001242796.1 Intron NP_001229725.1
NM_014669.4 Intron NP_055484.3
XM_005256263.3 Intron XP_005256320.1

View Full Product Details