Product Details

SNP ID
rs2353605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:225749 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTCATTTGTTAATGATGTTACTTA[C/T]TATTTGTTGATTTTCTTGAAGTTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC107986244 PubMed Links

Gene Details

Gene
LOC107986244
Gene Name
zinc finger protein 141-like
There are no transcripts associated with this gene.

Gene
ZNF876P
Gene Name
zinc finger protein 876, pseudogene
There are no transcripts associated with this gene.

View Full Product Details