Product Details

SNP ID
rs6774653
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:194340832 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGCAGCTTTTGAGGGTGCTTTGCAA[A/G]GCATAAGGATGGCCTTGTTAGGCCG
Phenotype
MIM: 603104
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPN2 PubMed Links

Gene Details

Gene
CPN2
Gene Name
carboxypeptidase N subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080513.3 1961 UTR 3 NP_001073982.3
NM_001291988.1 1961 UTR 3 NP_001278917.1
XM_005269280.4 1961 UTR 3 XP_005269337.1

View Full Product Details