Product Details

SNP ID
rs4718177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:64991278 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGATTTTAGCAGTTATTTCTTTGAT[A/G]ACCTTTCCTTCGTCATCAAGTTCCA
Phenotype
MIM: 131170 MIM: 194624
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERV3-1 PubMed Links

Gene Details

Gene
ERV3-1
Gene Name
endogenous retrovirus group 3 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007253.3 2343 Silent Mutation GTC,GTT V583V NP_001007254.2
Gene
ZNF117
Gene Name
zinc finger protein 117
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015852.3 2343 Intron NP_056936.2

View Full Product Details