Product Details

SNP ID
rs7648564
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:184054198 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGCATAGCACCAAGGTAAGCACC[C/T]AGTCAGATACCTACCGTGTTTCTCA
Phenotype
MIM: 610121
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HTR3C PubMed Links

Gene Details

Gene
HTR3C
Gene Name
5-hydroxytryptamine receptor 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130770.2 Intron NP_570126.2

View Full Product Details