Product Details
- SNP ID
-
rs4072738
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:17981233 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTGGGGCATCTTTGCTTCCTCCAGG[A/C]AAAACAGCAGATGAACCCAAAGGGG
- Phenotype
-
MIM: 615519
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
DRC3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77521760] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DRC3
- Gene Name
- dynein regulatory complex subunit 3
- Gene
- TOM1L2
- Gene Name
- target of myb1 like 2 membrane trafficking protein
There are no transcripts associated with this gene.
View Full Product Details