Product Details

SNP ID
rs4859584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:76021984 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAAGGGGATATAAAAGTGTGATA[C/G]TCTGACTCTGGCTTCAGATTGGGTT
Phenotype
MIM: 603086 MIM: 147310
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ART3 PubMed Links

Gene Details

Gene
ART3
Gene Name
ADP-ribosyltransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130016.2 Intron NP_001123488.1
NM_001130017.2 Intron NP_001123489.1
NM_001179.5 Intron NP_001170.2
XM_005262997.1 Intron XP_005263054.1
XM_005262999.1 Intron XP_005263056.1
XM_005263003.1 Intron XP_005263060.1
XM_005263004.1 Intron XP_005263061.1
XM_006714220.1 Intron XP_006714283.1
XM_011531971.1 Intron XP_011530273.1
XM_017008206.1 Intron XP_016863695.1
XM_017008207.1 Intron XP_016863696.1
XM_017008208.1 Intron XP_016863697.1
XM_017008209.1 Intron XP_016863698.1
XM_017008210.1 Intron XP_016863699.1
Gene
CXCL10
Gene Name
C-X-C motif chemokine ligand 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001565.3 Intron NP_001556.2

View Full Product Details