Product Details

SNP ID
rs7097775
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:5393955 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGCTGGTTGGAGGACTCAAAGCA[A/G]GCAGTGGTGATGTCTGACACAGAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TUBAL3 PubMed Links

Gene Details

Gene
TUBAL3
Gene Name
tubulin alpha like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171864.1 941 Silent Mutation GCC,GCT A261A NP_001165335.1
NM_024803.2 941 Silent Mutation GCC,GCT A301A NP_079079.1

View Full Product Details