Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000442.4 | 3963 | UTR 3 | NP_000433.4 | ||
XM_005276880.1 | 3963 | UTR 3 | XP_005276937.1 | ||
XM_005276881.1 | 3963 | UTR 3 | XP_005276938.1 | ||
XM_005276882.1 | 3963 | UTR 3 | XP_005276939.1 | ||
XM_005276883.2 | 3963 | Intron | XP_005276940.1 | ||
XM_011524889.2 | 3963 | UTR 3 | XP_011523191.1 | ||
XM_011524890.1 | 3963 | UTR 3 | XP_011523192.1 | ||
XM_017024738.1 | 3963 | UTR 3 | XP_016880227.1 | ||
XM_017024739.1 | 3963 | UTR 3 | XP_016880228.1 | ||
XM_017024740.1 | 3963 | UTR 3 | XP_016880229.1 | ||
XM_017024741.1 | 3963 | UTR 3 | XP_016880230.1 |