Product Details
- SNP ID
-
rs12008310
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:151569505 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTACTTTAAGGTGAAATTCACAAA[A/G]TCAATCAACATGAAGAATCTGGAAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PASD1
PubMed Links
Gene Details
- Gene
- PASD1
- Gene Name
- PAS domain containing 1
View Full Product Details