Product Details

SNP ID
rs12014640
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:85294035 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGGAGGAGAGGAACATTTAAAAA[G/T]AATTAAAGAAAATGCTACTGAATTT
Phenotype
MIM: 300603
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
POF1B PubMed Links

Gene Details

Gene
POF1B
Gene Name
premature ovarian failure, 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307940.1 Intron NP_001294869.1
NM_024921.3 Intron NP_079197.3
XM_005262203.3 Intron XP_005262260.1

View Full Product Details