Product Details
- SNP ID
-
rs12658147
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:171389537 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACATAATTTCTGAATATAGTTTGAG[A/G]CGTATGTCACTTTCTGTTGCTTTAA
- Phenotype
-
MIM: 164040
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR3912
PubMed Links
Gene Details
- Gene
- MIR3912
- Gene Name
- microRNA 3912
There are no transcripts associated with this gene.
- Gene
- NPM1
- Gene Name
- nucleophosmin
View Full Product Details