Product Details
- SNP ID
-
rs34198089
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:74843872 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGCCATGCCACTTCTCAGGACGTT[A/G]GCTTTTTTTTTTTTTTTTAGAAACA
- Phenotype
-
MIM: 138254
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GRIN2C
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs551372458] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GRIN2C
- Gene Name
- glutamate ionotropic receptor NMDA type subunit 2C
- Gene
- TMEM104
- Gene Name
- transmembrane protein 104
There are no transcripts associated with this gene.
View Full Product Details