Product Details

SNP ID
rs232219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:88865062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGAGACTCTGCCAGGATCCACC[G/A]TTCTACAAAATATCTGTGATGGACA
Phenotype
MIM: 147200
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
IGK PubMed Links

Gene Details

Gene
IGK
Gene Name
immunoglobulin kappa locus
There are no transcripts associated with this gene.

Gene
IGKC
Gene Name
immunoglobulin kappa constant
There are no transcripts associated with this gene.

View Full Product Details