Product Details

SNP ID
rs164087
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141954721 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGACAACTCTACAAGGTAGATAC[C/T]CTTACTATCATCTCTAATTTACAGA
Phenotype
MIM: 605622 MIM: 605675
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PCDH12 PubMed Links

Gene Details

Gene
PCDH12
Gene Name
protocadherin 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016580.3 Intron NP_057664.1
Gene
RNF14
Gene Name
ring finger protein 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201365.1 Intron NP_001188294.1
NM_004290.4 Intron NP_004281.1
NM_183398.2 Intron NP_899645.1
NM_183399.2 Intron NP_899646.1
NM_183400.2 Intron NP_899647.1
NM_183401.2 Intron NP_899648.1
XM_005268536.2 Intron XP_005268593.1
XM_005268537.2 Intron XP_005268594.1
XM_005268539.2 Intron XP_005268596.1
XM_005268540.3 Intron XP_005268597.1
XM_005268541.3 Intron XP_005268598.1
XM_005268542.2 Intron XP_005268599.1
XM_005268543.4 Intron XP_005268600.1
XM_011537714.2 Intron XP_011536016.1
XM_017010070.1 Intron XP_016865559.1
XM_017010071.1 Intron XP_016865560.1
XM_017010072.1 Intron XP_016865561.1
XM_017010073.1 Intron XP_016865562.1
XM_017010074.1 Intron XP_016865563.1
XM_017010075.1 Intron XP_016865564.1
XM_017010076.1 Intron XP_016865565.1
XM_017010077.1 Intron XP_016865566.1

View Full Product Details