Product Details
- SNP ID
-
rs684970
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
41
- Location
-
Chr.1:236397534 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGCAACATGGAACAAAGGGATTTC[A/G]AAATCCCAGGACATGTGGAGGAGTG
- Phenotype
-
MIM: 606603
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EDARADD
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76959088] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EDARADD
- Gene Name
- EDAR associated death domain
View Full Product Details