Product Details

SNP ID
rs1135216
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.6:32847198 on Build GRCh38
Set Membership
HapMap DME Validated Inventoried
Context Sequence [VIC/FAM]
CCCTGACAGCTGGCTCCCAGCCTCG[C/T]CTACCTCTGCAGAGCAAAGGGCCAA
Phenotype
MIM: 177046 MIM: 177045 MIM: 170260 MIM: 170261
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSMB8 PubMed Links

Gene Details

Gene
PSMB8
Gene Name
proteasome subunit beta 8
There are no transcripts associated with this gene.

Gene
PSMB8-AS1
Gene Name
PSMB8 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
PSMB9
Gene Name
proteasome subunit beta 9
There are no transcripts associated with this gene.

Gene
TAP1
Gene Name
transporter 1, ATP binding cassette subfamily B member
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000593.5 2245 Missense Mutation GAC,GGC D697G NP_000584.2
NM_001292022.1 2245 Missense Mutation GAC,GGC D436G NP_001278951.1
Gene
TAP2
Gene Name
transporter 2, ATP binding cassette subfamily B member
There are no transcripts associated with this gene.

View Full Product Details