Product Details

SNP ID
rs495580
Assay Type
Functionally tested
NCBI dbSNP Submissions
51
Location
Chr.1:93094622 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTGCATCCCCACTGTGTAGTTGA[A/G]CATATTTCTCTGCATTTACTAGATC
Phenotype
MIM: 609882
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTF2 PubMed Links

Gene Details

Gene
MTF2
Gene Name
metal response element binding transcription factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164391.1 Intron NP_001157863.1
NM_001164392.1 Intron NP_001157864.1
NM_001164393.1 Intron NP_001157865.1
NM_007358.3 Intron NP_031384.1
XM_011541016.1 Intron XP_011539318.1
XM_011541017.2 Intron XP_011539319.1
XM_011541019.1 Intron XP_011539321.1
XM_017000676.1 Intron XP_016856165.1
XM_017000677.1 Intron XP_016856166.1
XM_017000678.1 Intron XP_016856167.1
XM_017000679.1 Intron XP_016856168.1

View Full Product Details