Product Details

SNP ID
rs764763
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:78856706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACTGCTCCAGCTGTGCCTGGCA[A/C]AACCTCCGGGCTCATTTTAAGAGGA
Phenotype
MIM: 188825
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TIMP2 PubMed Links

Gene Details

Gene
TIMP2
Gene Name
TIMP metallopeptidase inhibitor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003255.4 Intron NP_003246.1

View Full Product Details