Product Details

SNP ID
rs380286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:1320132 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGAAGGCCAAGCTGAGAGGTCCAC[A/G]CAATGCAGTCTTCAGAGGCCCTGGT
Phenotype
MIM: 612585
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLPTM1L PubMed Links

Gene Details

Gene
CLPTM1L
Gene Name
CLPTM1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030782.4 Intron NP_110409.2
XM_011514144.2 Intron XP_011512446.1

View Full Product Details