Product Details

SNP ID
rs375171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:44077102 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTAAACAGATTGGTGGGGCCCTGG[A/G]GATAGAAGGAGAGAAAGTATATGTG
Phenotype
MIM: 611045 MIM: 605315 MIM: 611793
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
G6PC3 PubMed Links

Gene Details

Gene
G6PC3
Gene Name
glucose-6-phosphatase catalytic subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319945.1 4943 Intron NP_001306874.1
NM_138387.3 4943 Intron NP_612396.1
XM_011525473.2 4943 Intron XP_011523775.1
XM_011525474.2 4943 Intron XP_011523776.1
XM_017025335.1 4943 Intron XP_016880824.1
Gene
HDAC5
Gene Name
histone deacetylase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001015053.1 4943 UTR 3 NP_001015053.1
NM_005474.4 4943 UTR 3 NP_005465.2
XM_005256905.1 4943 UTR 3 XP_005256962.1
XM_005256906.4 4943 UTR 3 XP_005256963.1
XM_011524149.1 4943 UTR 3 XP_011522451.1
XM_011524150.1 4943 UTR 3 XP_011522452.1
XM_017023988.1 4943 UTR 3 XP_016879477.1
XM_017023989.1 4943 UTR 3 XP_016879478.1
XM_017023990.1 4943 UTR 3 XP_016879479.1
XM_017023991.1 4943 UTR 3 XP_016879480.1
XM_017023992.1 4943 UTR 3 XP_016879481.1
XM_017023993.1 4943 UTR 3 XP_016879482.1
XM_017023994.1 4943 Intron XP_016879483.1
XM_017023995.1 4943 UTR 3 XP_016879484.1
XM_017023996.1 4943 UTR 3 XP_016879485.1
Gene
LSM12
Gene Name
LSM12 homolog
There are no transcripts associated with this gene.

View Full Product Details