Product Details

SNP ID
rs177255
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:233838768 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCATCTGTTAAACAGAACGCATAAA[C/T]TTTGCCTCAAAGCAGCCTCCATGAA
Phenotype
MIM: 612667
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HJURP PubMed Links

Gene Details

Gene
HJURP
Gene Name
Holliday junction recognition protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282962.1 Intron NP_001269891.1
NM_001282963.1 Intron NP_001269892.1
NM_018410.4 Intron NP_060880.3
XM_011511437.1 Intron XP_011509739.1
Gene
MROH2A
Gene Name
maestro heat like repeat family member 2A
There are no transcripts associated with this gene.

View Full Product Details