Product Details

SNP ID
rs221939
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103608180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTGAGGAGGGGGAAGCGGAGCGG[G/T]CGGAGGCAAGGGCGGGGGAGCCTGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TCEAL3 PubMed Links

Gene Details

Gene
TCEAL3
Gene Name
transcription elongation factor A like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006933.1 Intron NP_001006934.1
NM_032926.2 Intron NP_116315.1

View Full Product Details