Product Details

SNP ID
rs726703
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:12384229 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTTGAGTCTTTCCTATTTGGAGAAA[A/G]TCTCCTGATACTGGTTTCCTCTTTA
Phenotype
MIM: 608120
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PARVA PubMed Links

Gene Details

Gene
PARVA
Gene Name
parvin alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018222.4 Intron NP_060692.2
XM_005253015.3 Intron XP_005253072.1

View Full Product Details