Product Details

SNP ID
rs14008
Assay Type
Functionally tested
NCBI dbSNP Submissions
85
Location
Chr.1:58576509 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
AGTAGGCGGCATCGCCGATATCCAC[G/T]TCACCGGCGGCCTTCTGAGACGTGT
Phenotype
MIM: 137290
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TACSTD2 PubMed Links

Gene Details

Gene
TACSTD2
Gene Name
tumor-associated calcium signal transducer 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002353.2 986 Silent Mutation NP_002344.2

View Full Product Details