Product Details

SNP ID
rs575147
Assay Type
Validated
NCBI dbSNP Submissions
54
Location
Chr.1:169923143 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CAGGTTAGCATTTGTACTGAGCGAT[C/T]CTGGACAAAATTTGTTGTTTTGGGG
Phenotype
MIM: 601836
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KIFAP3 PubMed Links
Additional Information
For this assay, SNP(s) [rs80175861] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KIFAP3
Gene Name
kinesin associated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204514.1 Intron NP_001191443.1
NM_001204516.1 Intron NP_001191445.1
NM_001204517.1 Intron NP_001191446.1
NM_014970.3 Intron NP_055785.2
XM_005244970.1 Intron XP_005245027.1
XM_011509307.2 Intron XP_011507609.2

View Full Product Details