Product Details
- SNP ID
-
rs310655
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:63540928 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCAGCATGAGCACGTAGACCTCCG[C/T]CGGGCAGGCAGCCGGGCGCGGCAGC
- Phenotype
-
MIM: 602004
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PTK6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs8120713] are located under a probe and SNP(s) [rs310654] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PTK6
- Gene Name
- protein tyrosine kinase 6
There are no transcripts associated with this gene.
- Gene
- SRMS
- Gene Name
- src-related kinase lacking C-terminal regulatory tyrosine and N-terminal myristylation sites
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080823.3 |
1398 |
Missense Mutation |
ACG,GCG |
T453A |
NP_543013.1 |
View Full Product Details