Product Details

SNP ID
rs229890
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:74302841 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCAAGGCCCTATCCCCACGCTGGCT[A/G]TCAGCTGTCCCCACTATCTGGATGG
Phenotype
MIM: 603432
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CLIP2 PubMed Links

Gene Details

Gene
CLIP2
Gene Name
CAP-Gly domain containing linker protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003388.4 Intron NP_003379.3
NM_032421.2 Intron NP_115797.1

View Full Product Details