Product Details

SNP ID
rs295443
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:47296211 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACACATTGGTCCTACTCCCCTTAC[C/T]GGTCTGAGCAGGATGTAGATGGTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL18 PubMed Links

Gene Details

Gene
KLHL18
Gene Name
kelch like family member 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025010.4 Intron NP_079286.2
XM_005265001.3 Intron XP_005265058.2
XM_005265002.4 Intron XP_005265059.2
XM_005265003.3 Intron XP_005265060.2
XM_006713079.1 Intron XP_006713142.1
XM_017006033.1 Intron XP_016861522.1
XM_017006034.1 Intron XP_016861523.1
XM_017006035.1 Intron XP_016861524.1

View Full Product Details