Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_152315.3 | 5064 | Intron | NP_689528.2 | ||
XM_011542594.2 | 5064 | UTR 3 | XP_011540896.1 | ||
XM_011542595.2 | 5064 | UTR 3 | XP_011540897.1 | ||
XM_011542596.2 | 5064 | UTR 3 | XP_011540898.1 | ||
XM_011542597.2 | 5064 | UTR 3 | XP_011540899.1 | ||
XM_011542598.2 | 5064 | UTR 3 | XP_011540900.1 | ||
XM_011542599.2 | 5064 | UTR 3 | XP_011540901.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_182495.5 | 5064 | Intron | NP_872301.2 | ||
XM_011542604.2 | 5064 | Intron | XP_011540906.1 | ||
XM_017017206.1 | 5064 | Intron | XP_016872695.1 | ||
XM_017017207.1 | 5064 | Intron | XP_016872696.1 | ||
XM_017017208.1 | 5064 | Intron | XP_016872697.1 | ||
XM_017017209.1 | 5064 | Intron | XP_016872698.1 | ||
XM_017017210.1 | 5064 | Intron | XP_016872699.1 | ||
XM_017017211.1 | 5064 | Intron | XP_016872700.1 | ||
XM_017017212.1 | 5064 | Intron | XP_016872701.1 |