Product Details

SNP ID
rs568055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:120543244 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACAGGTCTGCAGACAGGTACTTT[A/T]GTATAGTTATATCAGAATCTTAGAG
Phenotype
MIM: 615998
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RNF10 PubMed Links

Gene Details

Gene
RNF10
Gene Name
ring finger protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014868.4 Intron NP_055683.3
XM_005254013.2 Intron XP_005254070.1
XM_006719717.2 Intron XP_006719780.1
XM_017020282.1 Intron XP_016875771.1
XM_017020283.1 Intron XP_016875772.1

View Full Product Details