Product Details

SNP ID
rs686546
Assay Type
Validated
NCBI dbSNP Submissions
68
Location
Chr.1:23383684 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
GTGGCCCAGACATCACATGGTGCAC[A/G]TGCAAGTCAGAGAGCAGTTGGAAAC
Phenotype
MIM: 604128
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCEA3 PubMed Links

Gene Details

Gene
TCEA3
Gene Name
transcription elongation factor A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003196.2 1542 Intron NP_003187.1
XM_006710864.1 1542 UTR 3 XP_006710927.1
XM_011542053.2 1542 Intron XP_011540355.1
XM_017002200.1 1542 Missense Mutation ACG,ATG T484M XP_016857689.1
XM_017002201.1 1542 Missense Mutation ACG,ATG T463M XP_016857690.1
XM_017002202.1 1542 Missense Mutation ACG,ATG T463M XP_016857691.1
XM_017002203.1 1542 Missense Mutation ACG,ATG T447M XP_016857692.1
XM_017002204.1 1542 Intron XP_016857693.1

View Full Product Details