Product Details

SNP ID
rs754984
Assay Type
Validated
NCBI dbSNP Submissions
35
Location
Chr.1:23371627 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
CCAATTTAGGCTTTCAACATTATCT[C/G]TAAAGAAGGTTATACATTATGTCGG
Phenotype
MIM: 604128 MIM: 611703
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TCEA3 PubMed Links

Gene Details

Gene
TCEA3
Gene Name
transcription elongation factor A3
There are no transcripts associated with this gene.

Gene
ZNF436
Gene Name
zinc finger protein 436
There are no transcripts associated with this gene.

Gene
ZNF436-AS1
Gene Name
ZNF436 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details