Product Details

SNP ID
rs308935
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:77723984 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTCCCACAGAAAAAGTAAAATCA[A/G]CTGTTCCTTTTCTAGGGCTGCCATA
Phenotype
MIM: 609231
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUDT7 PubMed Links

Gene Details

Gene
NUDT7
Gene Name
nudix hydrolase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105663.2 Intron NP_001099133.1
NM_001243657.1 Intron NP_001230586.1
NM_001243660.1 Intron NP_001230589.1
NM_001243661.1 Intron NP_001230590.1
XM_011523032.2 Intron XP_011521334.1
XM_011523033.2 Intron XP_011521335.1
XM_017023170.1 Intron XP_016878659.1

View Full Product Details