Product Details

SNP ID
rs287780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:44146976 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATGGCTTAGTTATAAAAAGAAT[A/G]ATTATTTAAAGTGTCTGTATGGCTG
Phenotype
MIM: 300817
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EFHC2 PubMed Links

Gene Details

Gene
EFHC2
Gene Name
EF-hand domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025184.3 3872 Intron NP_079460.2
XM_006724562.2 3872 UTR 3 XP_006724625.1

View Full Product Details