Product Details
- SNP ID
-
rs439738
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:11280891 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- CAGGCCCAGAGACTAAGCCAAAGAC[C/G]TACTGCTATTAGCTCCTTTCTAATT
- Phenotype
-
MIM: 608760
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ATG7
PubMed Links
Gene Details
- Gene
- ATG7
- Gene Name
- autophagy related 7
View Full Product Details