Product Details

SNP ID
rs359468
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:173892055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGTGCTACAGCTAGAGGGTTAC[C/T]TTAGAAATATTTCTGCAAAGGCTGA
Phenotype
MIM: 610607
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPEB4 PubMed Links

Gene Details

Gene
CPEB4
Gene Name
cytoplasmic polyadenylation element binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308189.1 Intron NP_001295118.1
NM_001308191.1 Intron NP_001295120.1
NM_001308192.1 Intron NP_001295121.1
NM_001308193.1 Intron NP_001295122.1
NM_030627.3 Intron NP_085130.2
XM_005265994.1 Intron XP_005266051.1
XM_011534660.2 Intron XP_011532962.1
XM_011534661.2 Intron XP_011532963.1

View Full Product Details