Product Details
- SNP ID
-
rs162005
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:26867822 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTGACCAGGGATTAACTCTTCACCT[A/G]CCAGAGGTAGTTTTTCTTATTAAGT
- Phenotype
-
MIM: 600308
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AQP4
PubMed Links
Gene Details
- Gene
- AQP4
- Gene Name
- aquaporin 4
There are no transcripts associated with this gene.
- Gene
- AQP4-AS1
- Gene Name
- AQP4 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details