Product Details

SNP ID
rs41298442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:54844099 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCAACATTGTGCTGGTCACAGTT[T/C]TGCTGTTCATTTTCTGTACACCTCG
Phenotype
MIM: 600225
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
GCH1 PubMed Links

Gene Details

Gene
GCH1
Gene Name
GTP cyclohydrolase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000161.2 832 Missense Mutation AAA,AGA K224R NP_000152.1
NM_001024024.1 832 Missense Mutation AAA,AGA K224R NP_001019195.1
NM_001024070.1 832 Intron NP_001019241.1
NM_001024071.1 832 Intron NP_001019242.1
XM_017021218.1 832 Missense Mutation AAA,AGA K126R XP_016876707.1

View Full Product Details