Product Details
- SNP ID
-
rs343422
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:5189352 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAGGAAGCCTATAGACTAAACGGA[C/G]TTGGGTTTTTAAAATGGTGTGGTTA
- Phenotype
-
MIM: 616596
MIM: 607673
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ARL8B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78664388] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARL8B
- Gene Name
- ADP ribosylation factor like GTPase 8B
There are no transcripts associated with this gene.
- Gene
- EDEM1
- Gene Name
- ER degradation enhancing alpha-mannosidase like protein 1
View Full Product Details