Product Details

SNP ID
rs30479
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39312308 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGAAGATCTGACAGCCAGGAGCTG[A/G]GGCTCTCCACAGGTAGGGAAACAGC
Phenotype
MIM: 612807
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LRFN1 PubMed Links

Gene Details

Gene
LRFN1
Gene Name
leucine rich repeat and fibronectin type III domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020862.1 Intron NP_065913.1
XM_005259102.4 Intron XP_005259159.1
XM_017027033.1 Intron XP_016882522.1

View Full Product Details