Product Details

SNP ID
rs149277
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28554189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCGCGCCGCGGCTCCCGACCTCC[C/A]TGAGGCGGCGCTCTAGGGGCGGTGC
Phenotype
MIM: 613374
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
SGF29 PubMed Links

Gene Details

Gene
SGF29
Gene Name
SAGA complex associated factor 29
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138414.2 Intron NP_612423.1
XM_017022894.1 Intron XP_016878383.1

View Full Product Details