Product Details

SNP ID
rs182833
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:55880265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTGGCCGCACCGCCTCTGGGC[C/T]CCGGCGGCGAAGGCTCTTAGATGTC
Phenotype
MIM: 165120
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LYN PubMed Links

Gene Details

Gene
LYN
Gene Name
LYN proto-oncogene, Src family tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001111097.2 Intron NP_001104567.1
NM_002350.3 Intron NP_002341.1
XM_011517529.2 Intron XP_011515831.1
XM_017013415.1 Intron XP_016868904.1
XM_017013416.1 Intron XP_016868905.1

View Full Product Details