Product Details

SNP ID
rs752730
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:32869570 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTCCAGGTCATATTGGTAGGTGAC[A/G]TATTTAGCTCGCTCATTTAAGTGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KBTBD2 PubMed Links

Gene Details

Gene
KBTBD2
Gene Name
kelch repeat and BTB domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015483.2 2306 Silent Mutation TAC,TAT Y549Y NP_056298.2
XM_005249696.2 2306 Silent Mutation TAC,TAT Y549Y XP_005249753.1
XM_005249697.2 2306 Silent Mutation TAC,TAT Y491Y XP_005249754.1
XM_011515256.2 2306 Silent Mutation TAC,TAT Y549Y XP_011513558.1
XM_011515257.2 2306 Silent Mutation TAC,TAT Y491Y XP_011513559.1

View Full Product Details